Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6280MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: CAGB/426. Clonality: Monoclonal. Host: Mouse. Isotype: IgM kappa. Positive Control: Tonsil, lymph node, or spleen. Cellular Staining: Cytoplasmic. Buffer: 1mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CAGB protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 14kDa.Chromosome: 1q21. Gene Symbol: S100A9. Gene ID: 6280. Ensemble: ENSG00000163220. Uniprot: NP_002956.
Catalog No. 50-188-5153