Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S100A9 Human 250ug
SDP

Catalog No. 5000185046
Encompass_Preferred

Recombinant human S100A9 protein supplied as a lyophilized powder (250 μg). S100A9 is a calcium- and zinc-binding regulator involved in pro-inflammatory signaling, immune cell migration, and metal ion chelation; it forms a heterodimer with S100A8 and modulates NF-κB and STAT pathways. Intended for research use.

  • Recombinant human protein with UniProt accession P06702.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Expressed in E. coli and supplied tag free.
  • Available in multiple unit sizes; supplied lyophilized for stability.
  • Do not reconstitute below 100 μg/mL in ddH2O; aliquot and store frozen to avoid freeze-thaw.

Catalog No. 50-001-85046 Supplier Medchemexpress LLC Supplier No. HYP70532250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human S100A9 protein supplied as a lyophilized powder (250 μg). S100A9 is a calcium- and zinc-binding regulator involved in pro-inflammatory signaling, immune cell migration, and metal ion chelation; it forms a heterodimer with S100A8 and modulates NF-κB and STAT pathways. Intended for research use.

  • Recombinant human protein with UniProt accession P06702.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Expressed in E. coli and supplied tag free.
  • Available in multiple unit sizes; supplied lyophilized for stability.
  • Do not reconstitute below 100 μg/mL in ddH2O; aliquot and store frozen to avoid freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.