Learn More
Medchemexpress LLC S100A9 Human His 100ug
SDP
Catalog No. 5000185489
Encompass_Preferred
S100A9 Protein, Human (His) is a recombinant human calcium- and zinc-binding protein produced in E. coli and provided as a His-tagged, highly purified preparation for research applications. It is intended for use in biochemical, cell signaling, and immunological studies involving neutrophil chemotaxis, phagocytosis, and inflammatory pathway activation. Handle and store according to the supplier datasheet to maintain activity.
  • High purity (>95%) recombinant protein suitable for in vitro assays.
  • Low endotoxin level (<1 EU/μg) for cell-based studies.
  • Available in multiple sizes, including a 100 μg unit for convenient use.
  • Stable when stored at -20°C; aliquot and freeze for long-term storage.
  • Includes an N-terminal His tag to facilitate detection and purification.
Catalog No. 50-001-85489 Supplier Medchemexpress LLC Supplier No. HYP70532A100UG
S100A9 Protein, Human (His) is a recombinant human calcium- and zinc-binding protein produced in E. coli and provided as a His-tagged, highly purified preparation for research applications. It is intended for use in biochemical, cell signaling, and immunological studies involving neutrophil chemotaxis, phagocytosis, and inflammatory pathway activation. Handle and store according to the supplier datasheet to maintain activity.
  • High purity (>95%) recombinant protein suitable for in vitro assays.
  • Low endotoxin level (<1 EU/μg) for cell-based studies.
  • Available in multiple sizes, including a 100 μg unit for convenient use.
  • Stable when stored at -20°C; aliquot and freeze for long-term storage.
  • Includes an N-terminal His tag to facilitate detection and purification.