Learn More
Medchemexpress LLC S100A9 Human His 10ug
SDP
Catalog No. 5000184158
Encompass_Preferred
S100A9 Protein, Human (His) is a recombinant fragment of the human S100A9 calcium-binding protein supplied lyophilized for research use. The protein contains an N-terminal 6xHis affinity tag, is expressed in Escherichia coli, and is provided with recommended formulation and storage guidance for biochemical and immunological assays.
  • Recombinant human S100A9 with N-terminal His tag.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Supplied lyophilized with stabilizers to support storage.
  • Suitable for biochemical and immunological research applications.
  • Recommended reconstitution at 100 μg/mL or higher and aliquot storage at -20°C/-80°C.
Catalog No. 50-001-84158 Supplier Medchemexpress LLC Supplier No. HYP70532A10UG
S100A9 Protein, Human (His) is a recombinant fragment of the human S100A9 calcium-binding protein supplied lyophilized for research use. The protein contains an N-terminal 6xHis affinity tag, is expressed in Escherichia coli, and is provided with recommended formulation and storage guidance for biochemical and immunological assays.
  • Recombinant human S100A9 with N-terminal His tag.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Supplied lyophilized with stabilizers to support storage.
  • Suitable for biochemical and immunological research applications.
  • Recommended reconstitution at 100 μg/mL or higher and aliquot storage at -20°C/-80°C.