Learn More
Medchemexpress LLC S100A9 Human His 250ug
SDP
Catalog No. 5000184159
Encompass_Preferred
Recombinant human S100A9 (His) is a lyophilized, N-terminal 6xHis-tagged fragment of human S100A9 expressed in E. coli. Supplied with >95% purity and a defined amino acid sequence (T2-P114), it is suitable for biochemical characterization, receptor binding studies (TLR4/MD2, RAGE), structural analysis, and antibody development. Store frozen and follow reconstitution guidance to maintain activity.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Expressed in E. coli for consistent, scalable production.
  • Lyophilized formulation with stabilizers for long-term storage.
  • Supplied with a defined amino acid sequence (T2-P114).
  • Suitable for biochemical assays, receptor binding, and antibody generation.
  • Manufacturer recommends reconstitution to ≥100 μg/mL and aliquot freezing for storage.
Catalog No. 50-001-84159 Supplier Medchemexpress LLC Supplier No. HYP70532A250UG
Recombinant human S100A9 (His) is a lyophilized, N-terminal 6xHis-tagged fragment of human S100A9 expressed in E. coli. Supplied with >95% purity and a defined amino acid sequence (T2-P114), it is suitable for biochemical characterization, receptor binding studies (TLR4/MD2, RAGE), structural analysis, and antibody development. Store frozen and follow reconstitution guidance to maintain activity.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Expressed in E. coli for consistent, scalable production.
  • Lyophilized formulation with stabilizers for long-term storage.
  • Supplied with a defined amino acid sequence (T2-P114).
  • Suitable for biochemical assays, receptor binding, and antibody generation.
  • Manufacturer recommends reconstitution to ≥100 μg/mL and aliquot freezing for storage.