Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 6285MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: 4C4.9 + S100B/1012. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: Brain, Melanoma. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Purified bovine brain S100 protein (4C4.9) ;Recombinant full-length human S100B protein (S100B/1012). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 10-12kDa.Chromosome: 21q22.3. Gene Symbol: S100B. Gene ID: 6285. Ensemble: ENSG00000160307. Uniprot: XP_016883913.
Catalog No. 50-188-5159