Learn More
Sino Biological S100B Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10181T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human S100B (rh S100B; Catalog#10181-H07E; NP_006263.1; Ser2-Glu92). S100B specific IgG was purified by Human S100B affinity chromatography.
Catalog No. 50-224-6290