Learn More
ACROBiosystems Recombinant Protein;Human S100B Protein;E.coli;100UG;S1B,S100B,S100-B,NEF,S100,S100beta
Supplier:  ACROBiosystems S1BH5143100UG
Encompass_Preferred
Recombinant Protein;Human S100B Protein;100UG;Human S100B, His Tag (S1B-H5143) is expressed from E.coli cells. It contains AA Met 1 - Glu 92 (Accession # NP_006263).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 11.5 kDa. The protein migrates as 11 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8847