Learn More
Medchemexpress LLC S100P Human N-His 100ug
SDP
Catalog No. 5000196428
Encompass_Preferred
Recombinant human S100P protein with an N-terminal 6xHis tag, expressed in E. coli and supplied as a lyophilized powder. Purified to >95% by reducing SDS-PAGE and supplied in a 100 μg package with formulation designed for stability. The product is intended for in vitro research applications and is accompanied by a datasheet, COA, and SDS for batch-specific information.
  • High purity (>95%) for reliable assay performance.
  • N-terminal 6xHis tag for straightforward purification and detection.
  • Lyophilized formulation with trehalose for extended shelf life.
  • Low endotoxin (<1 EU/μg) suitable for sensitive assays.
  • Supplied with datasheet, COA, and SDS for batch-specific validation.
  • Recommended storage and reconstitution guidance to preserve activity.
Catalog No. 50-001-96428 Supplier Medchemexpress LLC Supplier No. HYP71138A100UG
Recombinant human S100P protein with an N-terminal 6xHis tag, expressed in E. coli and supplied as a lyophilized powder. Purified to >95% by reducing SDS-PAGE and supplied in a 100 μg package with formulation designed for stability. The product is intended for in vitro research applications and is accompanied by a datasheet, COA, and SDS for batch-specific information.
  • High purity (>95%) for reliable assay performance.
  • N-terminal 6xHis tag for straightforward purification and detection.
  • Lyophilized formulation with trehalose for extended shelf life.
  • Low endotoxin (<1 EU/μg) suitable for sensitive assays.
  • Supplied with datasheet, COA, and SDS for batch-specific validation.
  • Recommended storage and reconstitution guidance to preserve activity.