Learn More
Medchemexpress LLC S100P Human N-His 10ug
SDP
Catalog No. 5000196429
Encompass_Preferred
Recombinant human S100P protein with an N-terminal 6xHis affinity tag, expressed in Escherichia coli and supplied in a 10 μg research-scale vial. The protein is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 9-12 kDa, and includes manufacturer storage and reconstitution guidance to maintain integrity and activity.
  • High purity (>95%) suitable for biochemical assays.
  • N-terminal His tag enables affinity purification and detection.
  • Expressed in E. coli for consistent recombinant yield.
  • Small research-friendly 10 μg quantity for pilot experiments.
  • Stable when stored at -20°C; after reconstitution stable at 4°C for one week.
  • Recommended reconstitution at ≥100 μg/mL and use of carrier protein for long-term storage.
Catalog No. 50-001-96429 Supplier Medchemexpress LLC Supplier No. HYP71138A10UG
Recombinant human S100P protein with an N-terminal 6xHis affinity tag, expressed in Escherichia coli and supplied in a 10 μg research-scale vial. The protein is >95% pure by reducing SDS-PAGE, has an approximate molecular weight of 9-12 kDa, and includes manufacturer storage and reconstitution guidance to maintain integrity and activity.
  • High purity (>95%) suitable for biochemical assays.
  • N-terminal His tag enables affinity purification and detection.
  • Expressed in E. coli for consistent recombinant yield.
  • Small research-friendly 10 μg quantity for pilot experiments.
  • Stable when stored at -20°C; after reconstitution stable at 4°C for one week.
  • Recommended reconstitution at ≥100 μg/mL and use of carrier protein for long-term storage.