Learn More
Medchemexpress LLC S100P Human N-His 500ug
SDP
Catalog No. 5000196430
Encompass_Preferred
S100P Protein, Human (N-His) is a recombinant human calcium-binding protein supplied as a lyophilized powder, suitable for biochemical and cell biology research into calcium signaling and protein-protein interactions. It is expressed in E. coli and provided with an affinity tag to facilitate purification.
  • Recombinant human calcium-binding protein for biochemical assays and interaction studies.
  • Expressed in E. coli and supplied with an N-terminal 6xHis affinity tag for purification.
  • Lyophilized powder format enables long-term storage and convenient transport.
  • High purity (≥95% by reducing SDS-PAGE) suitable for analytical and preparative use.
  • Small molecular weight (9-12 kDa) consistent with expected S100 family proteins.
  • Stable when stored at -20°C; reconstitute at ≥100 μg/mL and add carrier protein for long-term storage.
Catalog No. 50-001-96430 Supplier Medchemexpress LLC Supplier No. HYP71138A500UG
S100P Protein, Human (N-His) is a recombinant human calcium-binding protein supplied as a lyophilized powder, suitable for biochemical and cell biology research into calcium signaling and protein-protein interactions. It is expressed in E. coli and provided with an affinity tag to facilitate purification.
  • Recombinant human calcium-binding protein for biochemical assays and interaction studies.
  • Expressed in E. coli and supplied with an N-terminal 6xHis affinity tag for purification.
  • Lyophilized powder format enables long-term storage and convenient transport.
  • High purity (≥95% by reducing SDS-PAGE) suitable for analytical and preparative use.
  • Small molecular weight (9-12 kDa) consistent with expected S100 family proteins.
  • Stable when stored at -20°C; reconstitute at ≥100 μg/mL and add carrier protein for long-term storage.