Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S100P Human N-His 50ug
SDP

Catalog No. 5000196517
Encompass_Preferred

Recombinant human S100P protein with an N-terminal His tag supplied as lyophilized powder for use in biochemical assays, antibody binding experiments, and molecular studies. The product is formulated and shipped with stability guidance to preserve activity and minimize endotoxin.

  • Endotoxin level <1 EU/μg, determined by LAL method.
  • Molecular weight approximately 9-12 kDa.
  • N-terminal His tag for affinity purification and detection.
  • Lyophilized from buffered solution with 8% trehalose for stability.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O.
  • Store lyophilized at -20°C; aliquot and freeze at -20°C or -80°C for long-term storage.
  • Suitable for antibody binding assays and biochemical characterization.

Catalog No. 50-001-96517 Supplier Medchemexpress LLC Supplier No. HYP71138A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human S100P protein with an N-terminal His tag supplied as lyophilized powder for use in biochemical assays, antibody binding experiments, and molecular studies. The product is formulated and shipped with stability guidance to preserve activity and minimize endotoxin.

  • Endotoxin level <1 EU/μg, determined by LAL method.
  • Molecular weight approximately 9-12 kDa.
  • N-terminal His tag for affinity purification and detection.
  • Lyophilized from buffered solution with 8% trehalose for stability.
  • Not recommended to reconstitute below 100 μg/mL in ddH2O.
  • Store lyophilized at -20°C; aliquot and freeze at -20°C or -80°C for long-term storage.
  • Suitable for antibody binding assays and biochemical characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.