Learn More
Pyramex™ S1035 Face Shield
Catalog No. 19155505 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Certifications/Compliance:
ANSI Z87.1
Catalog No. Certifications/Compliance
19-155-505 ANSI Z87.1
1 options
This item is not returnable. View return policy
Catalog No. 19-155-505 Supplier Pyramex™ Supplier No. S1035
Add to Cart
Add to Cart
This item is not returnable. View return policy
Dark green tinted polyethylene face shield
  • Meets ANSI Z87
  • 8 in. x 15 in. | 0.040 in. thick
  • Does not contain IR resin
  • Universal slots will accommodate a wide variety of headgear
  • Shown with HGBR - sold separately
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Green
    For Use With (Equipment) HGBR
    Weight (English) 2.5 oz.
    Material Polyethylene