Learn More
Pyramex™ S1060 Face Shield
Catalog No. 19155507 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Certifications/Compliance:
ANSI Z87.1
Catalog No. Certifications/Compliance
19-155-507 ANSI Z87.1
1 options
This item is not returnable. View return policy
Catalog No. 19-155-507 Supplier Pyramex™ Supplier No. S1060
Add to Cart
Add to Cart
This item is not returnable. View return policy
Wire mesh face shield
  • Meets ANSI Z87.1 standards for wire mesh
  • 8 in. x 15.5 in.
  • Provides good airflow for comfort 
  • Does not accommodate the HHAAD hard hat adapter
  • Shown with HGBR - sold separately
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Tint Dark Gray
    For Use With (Equipment) HGBR
    Weight (English) 2.9 oz.
    Material Steel Mesh, Nylon