Learn More
Cayman Chemical (S)-10-hydroxy-Camptothecin, Purity: ≥98%, CAS: 19685-09-7, 100 mg
SDP
Catalog No. 502968740
Encompass_Preferred
An inhibitor of topoisomerase I that demonstrates less toxicity than its parent compound; demonstrates strong anti-tumor activity against a wide range of experimental tumors including L1210 leukemia cells (IC50 = 1.15 μM)
Catalog No. 50-296-8740 Supplier Cayman Chemical Supplier No. 14635100 MG
An inhibitor of topoisomerase I that demonstrates less toxicity than its parent compound; demonstrates strong anti-tumor activity against a wide range of experimental tumors including L1210 leukemia cells (IC50 = 1.15 μM)