Learn More
Cayman Chemical S10hydroxyCamptothecIn 50mg
SDP
Catalog No. 502972741
Encompass_Preferred
An inhibitor of topoisomerase I that demonstrates less toxicity than its parent compound; demonstrates strong anti-tumor activity against a wide range of experimental tumors including L1210 leukemia cells (IC50 = 1.15 μM)
Catalog No. 50-297-2741 Supplier Cayman Chemical Supplier No. 1463550 MG
An inhibitor of topoisomerase I that demonstrates less toxicity than its parent compound; demonstrates strong anti-tumor activity against a wide range of experimental tumors including L1210 leukemia cells (IC50 = 1.15 μM)