Learn More
Pyramex™ S1150 Face Shield
Catalog No. 19155509 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
IR/UV 5.0
Catalog No. Lens Tint
19-155-509 IR/UV 5.0
1 options
This item is not returnable. View return policy
Catalog No. 19-155-509 Supplier Pyramex™ Supplier No. S1150
Add to Cart
Add to Cart
This item is not returnable. View return policy
Tapered IR 5 Polycarbonate face shield
  • Meets ANSI Z87+
  • 8 in. x 15.5 in. | .078 in. thick
  • Provides 99% UVA/B/C protection
  • Universal slots will accommodate a wide variety of headgear
  • Shown with HGBR - sold separately
  • Specifications
    Lens Tint IR/UV 5.0
    For Use With (Equipment) HGBR
    Weight (English) 5.6 oz.
    Material Polycarbonate