Learn More
Electron Microscopy Sciences PS12 Micrometer Standard with Graticules Cert
SDP
Catalog No. 5029006
Encompass_Preferred
These stage micrometers are available with a calibration certificate that provides traceability for the precise calibration and confirmation of accuracy of optical measuring instruments, necessary under ISO provisions. The glass discs are mounted in stainless steel slides with engraved serial numbers. Each slide is supplied in a polished wooden storage case. A Certificate of Comparison from Pyser-SGI, Graticules Division compares the scale to a NPL calibrated in-house standards. A statemen is provided on the accuracy of the item with respect to these standards.
Catalog No. 50-290-06 Supplier Electron Microscopy Sciences Supplier No. 6804712GR
These stage micrometers are available with a calibration certificate that provides traceability for the precise calibration and confirmation of accuracy of optical measuring instruments, necessary under ISO provisions. The glass discs are mounted in stainless steel slides with engraved serial numbers. Each slide is supplied in a polished wooden storage case. A Certificate of Comparison from Pyser-SGI, Graticules Division compares the scale to a NPL calibrated in-house standards. A statemen is provided on the accuracy of the item with respect to these standards.