Learn More
Medchemexpress LLC HY-112818 5mg Medchemexpress, S130 CAS:1160852-22-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1128185MG
Encompass_Preferred
Medchemexpress, HY-112818 5mg S130 CAS:1160852-22-1 S130 is a high affinity, selective inhibitor of ATG4B (a major cysteine protease) with an IC 50 of 3.24 µM. S130 suppresses autophagy flux [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2440