Learn More
Enzo Life Sciences S14-95 (0.5mg)
SDP
Supplier:  Enzo Life Sciences ALX350299MC05
Encompass_Preferred
Potent inhibitor of cytokine-induced activation of STAT1α leading to the inhibition of inducible expression of proinflammatory enzymes (COX-2, iNOS (NOS II)) and cytokines (TNF-α). Purity: ≥95% (HPLC). Solubility: Soluble in methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8837