Learn More
AdipoGen S14161 (1 mg)
SDP
Supplier:  AdipoGen AGCR13516M001
Encompass_Preferred
Chemical. CAS: 883046-50-2. Formula: C17H14FNO4. MW: 315.3. Potent antileukemia and antimyeloma compound. Apoptosis inducer. Delays tumor growth. Cyclin D2 promoter transactivation inhibitor. Inhibitor of pan-Class I PI3K isoforms.
Catalog No. 50-205-3897