Learn More
ATCC S16; Schwann Cells; Rat (Rattus norvegicus)
Catalog No. 502381936
Encompass
S16 is derived from a primary culture of Schwann cells that was isolated in 1989 from the sciatic nerve of Rattus norvegicus (rat). It was immortalized by repetitive passaging and is useful for investigating the cell biology of MAG and other myelin-related components in neuroscience and toxicology research.
Catalog No. 50-238-1936 Supplier ATCC Supplier No. CRL2941
S16 is derived from a primary culture of Schwann cells that was isolated in 1989 from the sciatic nerve of Rattus norvegicus (rat). It was immortalized by repetitive passaging and is useful for investigating the cell biology of MAG and other myelin-related components in neuroscience and toxicology research.