Learn More
BRANSON ULTRASONICS CORPORATION (3P) S161212CAL ULTRASONIC BATHS
Catalog No. NC3721445
Encompass_Preferred
Branson Ultrasonics M Series Ultrasonic Cleaning Bath, Model: M8800-E, Voltage: 230/240V, Capacity: 20.8L (5.5 gal.), Dimensions Interior: 49.5 x 29 x 15cm (19.5 x 11.5 x 6 in.), Hertz: 40kHz, Includes: Tank drains with valves, Plastic Housing, Stainless Steel Tank, Plug Type: CEE 7/7
Catalog No. NC3721445 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. CPX952836R
Add to Cart
Add to Cart
Request a Quote
Branson Ultrasonics M Series Ultrasonic Cleaning Bath, Model: M8800-E, Voltage: 230/240V, Capacity: 20.8L (5.5 gal.), Dimensions Interior: 49.5 x 29 x 15cm (19.5 x 11.5 x 6 in.), Hertz: 40kHz, Includes: Tank drains with valves, Plastic Housing, Stainless Steel Tank, Plug Type: CEE 7/7