Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC S18-000003 | 2068119-11-7 | 99.3% | 100 MG
SDP

Catalog No. 5000234093
Encompass_Preferred

A potent, selective, orally active inhibitor of retinoic acid receptor-related orphan receptor-gamma-t (RORγt) intended for research use. Demonstrates sub-30 nM binding potency against human RORγt and high selectivity over other ROR family members.

  • Potent RORγt inhibitor with sub-30 nM binding potency.
  • High selectivity for RORγt over other ROR family members.
  • High purity (99.3%), characterized by COA and analytical data.
  • Molecular formula C26H25F3N2O4S; molecular weight 518.55 g·mol⁻¹.
  • Soluble in DMSO for assay and in vivo formulation use.
  • Available in multiple pack sizes, including 100 mg.
  • Includes datasheet, SDS, and COA documentation.

Catalog No. 50-002-34093 Supplier Medchemexpress LLC Supplier No. HY119366100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A potent, selective, orally active inhibitor of retinoic acid receptor-related orphan receptor-gamma-t (RORγt) intended for research use. Demonstrates sub-30 nM binding potency against human RORγt and high selectivity over other ROR family members.

  • Potent RORγt inhibitor with sub-30 nM binding potency.
  • High selectivity for RORγt over other ROR family members.
  • High purity (99.3%), characterized by COA and analytical data.
  • Molecular formula C26H25F3N2O4S; molecular weight 518.55 g·mol⁻¹.
  • Soluble in DMSO for assay and in vivo formulation use.
  • Available in multiple pack sizes, including 100 mg.
  • Includes datasheet, SDS, and COA documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.