Learn More
Medchemexpress LLC HY-119366 5mg Medchemexpress, S18-000003 CAS:2068119-11-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1193665MG
Encompass_Preferred
Medchemexpress, HY-119366 5mg S18-000003 CAS:2068119-11-7 S18-000003 is a potent, selective and orally active inhibitor of retinoic acid receptor-related orphan receptor-gamma-t (RORγt), with an IC50 of <30 nM towards human RORγt in competitive binding assays. S18-000003 shows selectivity for RORγt over other ROR family members (IC50s>10 μM). S18-000003 can be used for the research of psoriasis with low risk of thymic aberrations. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0927