Learn More
Cayman Chemical S1HtrIfluoroacetate salt 10mg
SDP
Catalog No. 502917384
Encompass_Preferred
A synthetic peptide antagonist of the human growth hormone receptor; decreases STAT5 phosphorylation induced by human growth hormone in SK-MEL-28 and MALME-3M cell lines, as well as by prolactin in IM-9 B cells, at 100 and 200 nM
Catalog No. 50-291-7384 Supplier Cayman Chemical Supplier No. 3751510 MG
A synthetic peptide antagonist of the human growth hormone receptor; decreases STAT5 phosphorylation induced by human growth hormone in SK-MEL-28 and MALME-3M cell lines, as well as by prolactin in IM-9 B cells, at 100 and 200 nM