Learn More
Echelon Biosciences Research Labs S1P BEADS 1 mL
SDP
Catalog No. 50720783
Encompass
S1P Beads are composed of agarose with 10 nanomoles of bound Sphingosine 1-Phosphate (S1P) per mL of beads, enough for several protein binding experiments. Each 1 mL order of S1P Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-783 Supplier Echelon Biosciences Research Labs Supplier No. S61101ML
S1P Beads are composed of agarose with 10 nanomoles of bound Sphingosine 1-Phosphate (S1P) per mL of beads, enough for several protein binding experiments. Each 1 mL order of S1P Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable