Learn More
Echelon Biosciences Research Labs S1P BIOTIN 50 ug
SDP
Supplier:  Echelon Biosciences Research Labs S200B50UG
Encompass
Sphingosine 1-phosphate (S1P) is a bioactive lysophospholipid formed by phosphorylation of sphingosine by sphingosine kinases. S1P binds to five G-protein coupled receptors designated S1P1-5 leading to cell proliferation, motility, and has a role in regulation cell shape. In addition, S1P acts as an intracellular mediator in a receptor-independent manner regulating cell survival and Ca2+ homeostasis. Sphingosine 1-phosphate biotin (S1P-biotin) is a S1P analog with natural erythro stereochemistry labeled at the terminus of the alkyl chain with biotin. This item is non returnable
Catalog No. 50-720-770