Learn More
Selleck Chemical LLC S1RA
SDP
Catalog No. 5000002896
Encompass_Preferred
S1RA (E-52862) is a selective sigma-1 receptor ( 1R) antagonist with a reported binding affinity of Ki 17 0 7 0 nM for human 1 receptor selective over the sigma-2 receptor and against a panel of other 170 receptors enzymes transporters and ion channels
Catalog No. 50-000-02896 Supplier Selleck Chemical LLC Supplier No. S67542MG
S1RA (E-52862) is a selective sigma-1 receptor ( 1R) antagonist with a reported binding affinity of Ki 17 0 7 0 nM for human 1 receptor selective over the sigma-2 receptor and against a panel of other 170 receptors enzymes transporters and ion channels