Learn More
Medchemexpress LLC S1ra (hydrochloride) | 1265917-14-3 | 99.9% | 373.88 | 250 MG
SDP
Catalog No. 5000384784
Encompass_Preferred
S1RA (E-52862) hydrochloride is an orally active and selective sigma-1 receptor (σ1R) antagonist with a Ki value of 17 nM. It demonstrates good selectivity against σ2R (Ki >1000 nM). This compound is also identified as a human 5-HT2B receptor antagonist with an IC50 value of 4.7 μM. S1RA hydrochloride has been shown to inhibit neuropathic pain and activity-induced spinal sensitization.
  • Orally active and selective sigma-1 receptor (σ1R) antagonist
  • High affinity for human and guinea pig σ1Rs
  • Good selectivity against sigma-2 receptors (σ2R)
  • Acts as a human 5-HT2B receptor antagonist
  • Inhibits neuropathic pain
  • Reduces activity-induced spinal sensitization
  • Demonstrates analgesic effects on formalin-evoked nociceptive behaviors in mice
  • Reverses capsaicin-induced mechanical hypersensitivity in mice
Catalog No. 50-003-84784 Supplier Medchemexpress LLC Supplier No. 250MGHY18099A
S1RA (E-52862) hydrochloride is an orally active and selective sigma-1 receptor (σ1R) antagonist with a Ki value of 17 nM. It demonstrates good selectivity against σ2R (Ki >1000 nM). This compound is also identified as a human 5-HT2B receptor antagonist with an IC50 value of 4.7 μM. S1RA hydrochloride has been shown to inhibit neuropathic pain and activity-induced spinal sensitization.
  • Orally active and selective sigma-1 receptor (σ1R) antagonist
  • High affinity for human and guinea pig σ1Rs
  • Good selectivity against sigma-2 receptors (σ2R)
  • Acts as a human 5-HT2B receptor antagonist
  • Inhibits neuropathic pain
  • Reduces activity-induced spinal sensitization
  • Demonstrates analgesic effects on formalin-evoked nociceptive behaviors in mice
  • Reverses capsaicin-induced mechanical hypersensitivity in mice