Learn More
Medchemexpress LLC S2-16 | 672333-56-1 | 97.4% | 2973.36 | 1 MG
SDP
Catalog No. 5000423683
Encompass_Preferred
S2-16 is a synthetic peptide derived from the S2 region of cardiac myosin. It functions as a cryptic epitope, capable of inducing myocarditis in Lewis rats. A cryptic epitope is defined as an epitope that remains unrecognized by antibodies or T cells after an animal has been immunized with the intact antigen. This characteristic makes S2-16 a valuable tool for investigating the underlying mechanisms of autoimmune myocarditis.
  • Synthetic peptide from the S2 region of cardiac myosin.
  • Induces myocarditis in Lewis rats.
  • A cryptic epitope, meaning it is not recognized by antibodies or T cells after immunization with the intact antigen.
  • Useful for studying the mechanism of autoimmune myocarditis.
Catalog No. 50-004-23683 Supplier Medchemexpress LLC Supplier No. HYP105031MG
S2-16 is a synthetic peptide derived from the S2 region of cardiac myosin. It functions as a cryptic epitope, capable of inducing myocarditis in Lewis rats. A cryptic epitope is defined as an epitope that remains unrecognized by antibodies or T cells after an animal has been immunized with the intact antigen. This characteristic makes S2-16 a valuable tool for investigating the underlying mechanisms of autoimmune myocarditis.
  • Synthetic peptide from the S2 region of cardiac myosin.
  • Induces myocarditis in Lewis rats.
  • A cryptic epitope, meaning it is not recognized by antibodies or T cells after immunization with the intact antigen.
  • Useful for studying the mechanism of autoimmune myocarditis.