Learn More
Medchemexpress LLC S2-16 | 672333-56-1 | 97.4% | 2973.36 | 5 MG
SDP
Catalog No. 5000423229
Encompass_Preferred
S2-16 is a synthetic peptide derived from the S2 region of cardiac myosin. It functions as a cryptic epitope, meaning it's not typically recognized by antibodies or T cells when the animal is immunized with the intact antigen. S2-16 has been shown to induce myocarditis in Lewis rats and can be utilized to study the mechanism of autoimmune myocarditis.
  • For research use only
  • Not sold to patients
  • Purity: 97.41%
Catalog No. 50-004-23229 Supplier Medchemexpress LLC Supplier No. HYP105035MG
S2-16 is a synthetic peptide derived from the S2 region of cardiac myosin. It functions as a cryptic epitope, meaning it's not typically recognized by antibodies or T cells when the animal is immunized with the intact antigen. S2-16 has been shown to induce myocarditis in Lewis rats and can be utilized to study the mechanism of autoimmune myocarditis.
  • For research use only
  • Not sold to patients
  • Purity: 97.41%