Learn More
Expression Systems S2 DROSOPHILA CELLS ESF 921
SDP
Catalog No. 501656614
Encompass_Preferred
Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
The S2 insect cell line is derived from a primary culture of late stage (20-24 hours old) Drosophila melanogaster embryos. S2 cells are adapted for suspension culture in ESF 921 and are available as a frozen vial or suspension culture.
• Recombinant protein expression from transient expression or stable cell line
• Good growth in adherent or suspension culture
• Quality and performance testing
To Download Instructions for Use and/or COA please visit: www.expressionsystems.com
Catalog No. 50-165-6614 Supplier Expression Systems Supplier No. 94005F
Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
The S2 insect cell line is derived from a primary culture of late stage (20-24 hours old) Drosophila melanogaster embryos. S2 cells are adapted for suspension culture in ESF 921 and are available as a frozen vial or suspension culture.
• Recombinant protein expression from transient expression or stable cell line
• Good growth in adherent or suspension culture
• Quality and performance testing
To Download Instructions for Use and/or COA please visit: www.expressionsystems.com