Learn More
Medchemexpress LLC S2116 | 2262489-89-2 | 98.1% | 437.91 | 50 MG
SDP
Catalog No. 5000351775
Encompass_Preferred
S2116 is an N-alkylated tranylcypromine (TCP) derivative and a potent lysine-specific demethylase 1 (LSD1) inhibitor. It enhances H3K9 methylation and reciprocal H3K27 deacetylation at super-enhancer regions. S2116 can induce apoptosis in TCP-resistant T-cell acute lymphoblastic leukemia (T-ALL) cells by suppressing the transcription of NOTCH3 and TAL1 genes and has been shown to significantly slow the growth of T-ALL cells in xenotransplanted mice.
  • Potent lysine-specific demethylase 1 (LSD1) inhibitor
  • Effective for T-ALL cell lines with IC50 values ranging from 1.1 μM to 6.8 μM
  • Induces apoptosis and down-regulates NOTCH3 and TAL1 protein expression in T-ALL cells
  • Significantly reduces subcutaneous tumor size in xenotransplanted mice
  • Exhibits favorable pharmacokinetic properties in mice with a T1/2 of 3.76 hours
Catalog No. 50-003-51775 Supplier Medchemexpress LLC Supplier No. HY13652250MG
S2116 is an N-alkylated tranylcypromine (TCP) derivative and a potent lysine-specific demethylase 1 (LSD1) inhibitor. It enhances H3K9 methylation and reciprocal H3K27 deacetylation at super-enhancer regions. S2116 can induce apoptosis in TCP-resistant T-cell acute lymphoblastic leukemia (T-ALL) cells by suppressing the transcription of NOTCH3 and TAL1 genes and has been shown to significantly slow the growth of T-ALL cells in xenotransplanted mice.
  • Potent lysine-specific demethylase 1 (LSD1) inhibitor
  • Effective for T-ALL cell lines with IC50 values ranging from 1.1 μM to 6.8 μM
  • Induces apoptosis and down-regulates NOTCH3 and TAL1 protein expression in T-ALL cells
  • Significantly reduces subcutaneous tumor size in xenotransplanted mice
  • Exhibits favorable pharmacokinetic properties in mice with a T1/2 of 3.76 hours