Learn More
Diapharma Group, Inc. S2288 SUBSTRATE
SDP
Catalog No. NC9894332
Encompass
Chromogenic substrate for t-PA and a broad spectrum of other serine proteases Package: 25 mg Molecular wt: 577.6 Formula: H-D-Ile-Pro-Arg-pNA -- 2HCl Solubility: >40mmol/L in H 2 O
Catalog No. NC9894332 Supplier Diapharma Group, Inc. Supplier No. S820852
Chromogenic substrate for t-PA and a broad spectrum of other serine proteases Package: 25 mg Molecular wt: 577.6 Formula: H-D-Ile-Pro-Arg-pNA -- 2HCl Solubility: >40mmol/L in H 2 O