Learn More
Medchemexpress LLC HY-122136 5mg Medchemexpress, S26131 CAS:296280-56-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1221365MG
Encompass_Preferred
Medchemexpress, HY-122136 5mg S26131 CAS:296280-56-3 S26131 (compound 5) is a potent and selective MT1 melatoninergic ligand, and the Ki values are 0.5 and 112 nM for MT1 and MT2, respectively. S26131 behaves as an MT1 and MT2 antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9627