Learn More
FUJIFILM Biosciences S262 Rat Mono AntIBODY 50uL
Catalog No. 5000450170
Encompass_Preferred
Tau is one type of microtubule-binding protein that is primarily expressed within the neurons of the central nervous system and is involved in the modulation of microtubule stability In Alzheimers disease the formation of neurofibrillary tangles with accumulated phosphorylated Tau proteins is known to occur Since correlations between the occurrence frequency of neurofibrillary tangles and severity of dementia were reported neurofibrillary tangles have been studied for determination of the causes and development of therapeutic agents for Alzheimers disease This product is a rat monoclonal antibody which recognizes Tau phosphorylated at the 262th serine Specific to Tau phosphorylated at the 262th serine
Catalog No. 50-004-50170 Supplier FUJIFILM Biosciences Supplier No. 01027123
Tau is one type of microtubule-binding protein that is primarily expressed within the neurons of the central nervous system and is involved in the modulation of microtubule stability In Alzheimers disease the formation of neurofibrillary tangles with accumulated phosphorylated Tau proteins is known to occur Since correlations between the occurrence frequency of neurofibrillary tangles and severity of dementia were reported neurofibrillary tangles have been studied for determination of the causes and development of therapeutic agents for Alzheimers disease This product is a rat monoclonal antibody which recognizes Tau phosphorylated at the 262th serine Specific to Tau phosphorylated at the 262th serine