Learn More
Electron Microscopy Sciences S29 Stage Counting Grid, 0.01mm Squares
SDP
Catalog No. 5031227
Encompass_Preferred
Stage graticules are used for the routine calibration of eyepiece patterns, particularly when alternating between objectives on one microscope or when using the same graticule in different microscopes. Their robust construction makes them ideal for student use and for instructional purposes. The scale or grid is centered on a glass disc mounted in a black anodized aluminum slide 75 mm x 24 mm x 2 mm thick. Transmitted light stage slide with 1.5mm x 1.5mm grid of 0.01mm squares
Catalog No. 50-312-27 Supplier Electron Microscopy Sciences Supplier No. 6804529
Stage graticules are used for the routine calibration of eyepiece patterns, particularly when alternating between objectives on one microscope or when using the same graticule in different microscopes. Their robust construction makes them ideal for student use and for instructional purposes. The scale or grid is centered on a glass disc mounted in a black anodized aluminum slide 75 mm x 24 mm x 2 mm thick. Transmitted light stage slide with 1.5mm x 1.5mm grid of 0.01mm squares