Learn More
Medchemexpress LLC HY-122614 5mg Medchemexpress, S29434 CAS:874484-20-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1226145MG
Encompass_Preferred
Medchemexpress, HY-122614 5mg S29434 CAS:874484-20-5 S29434 (NMDPEF) is a potent, competitive, selective and cell-permeable inhibitor of quinone reductase 2 (QR2), with IC50s ranging from 5 to 16 nM for human QR2 at different organizational levels, and has good selectivity for QR2 over QR1. S29434 (NMDPEF) induces autophagy and inhibits QR2-mediated ROS production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9633