Learn More
TARGETMOL CHEMICALS INC S3I-201 50MG
SDP
Catalog No. 502580771
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. S3I-201 (S3I-201) is a selective Stat3 inhibitor (IC50 86±33 uM) and low effect towards STAT1/5. purity: 99%
Catalog No. 50-258-0771 Supplier TARGETMOL CHEMICALS INC Supplier No. T250550MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. S3I-201 (S3I-201) is a selective Stat3 inhibitor (IC50 86±33 uM) and low effect towards STAT1/5. purity: 99%