Learn More
Waters Corp Spherisorb ODS2 Column, 80Å, 3 µm, 4.6 mm X 150 mm, 1/pk
Supplier:  Waters Corp PSS832113
Encompass_Preferred
Spherisorb ODS2 columns are general purpose, silica-based, reversed-phase C18 columns. The ODS2 packing features intermediate ligand density with a specified carbon load of 11.5%.
Catalog No. 50-818-769