Learn More
MSC Honeywell MTS Fit Safety Shoe, Composite Toe, S3S, 1 pr.
Catalog No. 5000507226
Encompass
The MTS Fit S3S is a non-metallic safety shoe designed for ergonomic efficiency and comfort in industrial environments. Utilizing high-tech materials and a sporty design, this unisex shoe provides comprehensive protection for use in logistics, services, warehousing, and transportation. It features a water-resistant, breathable upper and is engineered for all-day performance and stability.
  • Composite toe cap protects against 200 J impact and compression
  • Non-metallic, anti-perforation midsole
  • Dual-density injected PU outsole with SR slip resistance and oil resistance
  • Heat and cold insulated sole
  • ESD certified for electrical resistance
  • Modular three-width insock fitting system
  • TPU heel counter for stability and support
  • Flat elastic laces for a secure fit
  • Compliant with ASTM F2413-18, ASTM F3445-21, and EN ISO 20345:2022 standards
Catalog No. 50-005-07226 Supplier MSC Supplier No. 79458360
The MTS Fit S3S is a non-metallic safety shoe designed for ergonomic efficiency and comfort in industrial environments. Utilizing high-tech materials and a sporty design, this unisex shoe provides comprehensive protection for use in logistics, services, warehousing, and transportation. It features a water-resistant, breathable upper and is engineered for all-day performance and stability.
  • Composite toe cap protects against 200 J impact and compression
  • Non-metallic, anti-perforation midsole
  • Dual-density injected PU outsole with SR slip resistance and oil resistance
  • Heat and cold insulated sole
  • ESD certified for electrical resistance
  • Modular three-width insock fitting system
  • TPU heel counter for stability and support
  • Flat elastic laces for a secure fit
  • Compliant with ASTM F2413-18, ASTM F3445-21, and EN ISO 20345:2022 standards