Learn More
MSC MTS Fit S3S Sporty Workshoe, Composite Toe, Size 6, 1/Pr.
Catalog No. 5000507321
Encompass
These low-cut sporty work shoes are designed for performance and safety in environments such as logistics, warehouses, services, and transportation. Featuring a flexible, lightweight, and non-metallic construction, they provide robust protection while maintaining a casual aesthetic suitable for indoor and outdoor use.
  • Composite toe cap resistant to 200 Joules impact
  • Anti-perforation, non-metallic midsole
  • Dual-density injected polyurethane outsole with slip resistance, oil resistance, and electrical resistance (ESD)
  • Water-resistant and breathable upper construction
  • Thermoformed, antistatic, and anatomic insock with modular width fitting system
  • Heel energy absorption and insulation against heat and cold
  • Conforms to ASTM F2413-18, ASTM F3445-21, and EN ISO 20345:2022 standards
Catalog No. 50-005-07321 Supplier MSC Supplier No. 79451076
These low-cut sporty work shoes are designed for performance and safety in environments such as logistics, warehouses, services, and transportation. Featuring a flexible, lightweight, and non-metallic construction, they provide robust protection while maintaining a casual aesthetic suitable for indoor and outdoor use.
  • Composite toe cap resistant to 200 Joules impact
  • Anti-perforation, non-metallic midsole
  • Dual-density injected polyurethane outsole with slip resistance, oil resistance, and electrical resistance (ESD)
  • Water-resistant and breathable upper construction
  • Thermoformed, antistatic, and anatomic insock with modular width fitting system
  • Heel energy absorption and insulation against heat and cold
  • Conforms to ASTM F2413-18, ASTM F3445-21, and EN ISO 20345:2022 standards