Learn More
MSC Mts Sporty Fit Safety Shoe, Ondura Nilo, Size 9, 1/Pr.
Catalog No. 5000507230
Encompass
These low-cut, unisex sporty work shoes offer a lightweight, flexible solution for logistics, services, warehousing, and transportation environments. Designed with a casual athletic style, they feature a water-resistant Ondura Nilo upper and a breathable full Artico lining. The footwear incorporates a non-metallic composite toe cap and anti-perforation midsole to provide essential protection.
  • Composite toe cap protects against impact and compression
  • Dual-density injected PU outsole is oil and slip resistant, HI-CI insulated, and silicone-free
  • ESD and antistatic properties
  • Thermoformed, anatomic PU foam insole
  • TPU heel counter adds stability and support
  • Modular three-width insock fitting system allows a customized fit
  • Compliant with ASTM F2413-18, ASTM F3445-21, and EN ISO 20345:2022 standards
Catalog No. 50-005-07230 Supplier MSC Supplier No. 79454880
These low-cut, unisex sporty work shoes offer a lightweight, flexible solution for logistics, services, warehousing, and transportation environments. Designed with a casual athletic style, they feature a water-resistant Ondura Nilo upper and a breathable full Artico lining. The footwear incorporates a non-metallic composite toe cap and anti-perforation midsole to provide essential protection.
  • Composite toe cap protects against impact and compression
  • Dual-density injected PU outsole is oil and slip resistant, HI-CI insulated, and silicone-free
  • ESD and antistatic properties
  • Thermoformed, anatomic PU foam insole
  • TPU heel counter adds stability and support
  • Modular three-width insock fitting system allows a customized fit
  • Compliant with ASTM F2413-18, ASTM F3445-21, and EN ISO 20345:2022 standards