Learn More
Cayman Chemical S49076 (hydrochloride), Purity: ≥98%, CAS: 1265965-19-2, 10 mg
SDP
Catalog No. 502943708
Encompass_Preferred
A multi-kinase inhibitor; inhibits c-Met, Axl, Mer, FGFR1, FGFR2, and FGFR3 (IC50s = 1, 7, 2, 18, 17, and 15 nM, respectively); inhibits Axl- and c-Met-mediated Akt phosphorylation in MDA-MB-231 cells (IC50s = 33 and 50s = 50-200 nM); reduces mean tumor volume in GTL-16 and U87MG mouse xenograft models from 6.25-50 mg/kg
Catalog No. 50-294-3708 Supplier Cayman Chemical Supplier No. 4117310 MG
A multi-kinase inhibitor; inhibits c-Met, Axl, Mer, FGFR1, FGFR2, and FGFR3 (IC50s = 1, 7, 2, 18, 17, and 15 nM, respectively); inhibits Axl- and c-Met-mediated Akt phosphorylation in MDA-MB-231 cells (IC50s = 33 and 50s = 50-200 nM); reduces mean tumor volume in GTL-16 and U87MG mouse xenograft models from 6.25-50 mg/kg