Learn More
Electron Microscopy Sciences PS5 Micrometer Standard with NPL Cert
SDP
Catalog No. 5028996
Encompass_Preferred
These stage micrometers are available with a calibration certificate that provides traceability for the precise calibration and confirmation of accuracy of optical measuring instruments, necessary under ISO provisions. The glass discs are mounted in stainless steel slides with engraved serial numbers. Each slide is supplied in a polished wooden storage case. Calibrated by the National Physical Laboratory by taking measurements at selected points on the scale.
Catalog No. 50-289-96 Supplier Electron Microscopy Sciences Supplier No. 680475NP
These stage micrometers are available with a calibration certificate that provides traceability for the precise calibration and confirmation of accuracy of optical measuring instruments, necessary under ISO provisions. The glass discs are mounted in stainless steel slides with engraved serial numbers. Each slide is supplied in a polished wooden storage case. Calibrated by the National Physical Laboratory by taking measurements at selected points on the scale.