Learn More
Electron Microscopy Sciences PS5 Micrometer Standard with UKAS Cert
SDP
Catalog No. 5028995
Encompass_Preferred
These stage micrometers are available with a calibration certificate that provides traceability for the precise calibration and confirmation of accuracy of optical measuring instruments, necessary under ISO provisions. The glass discs are mounted in stainless steel slides with engraved serial numbers. Each slide is supplied in a polished wooden storage case. A UKAS accredited laboratory carries out measurements at selected points on the scales and grids and issues a calibration certificate.
Catalog No. 50-289-95 Supplier Electron Microscopy Sciences Supplier No. 680475NA
These stage micrometers are available with a calibration certificate that provides traceability for the precise calibration and confirmation of accuracy of optical measuring instruments, necessary under ISO provisions. The glass discs are mounted in stainless steel slides with engraved serial numbers. Each slide is supplied in a polished wooden storage case. A UKAS accredited laboratory carries out measurements at selected points on the scales and grids and issues a calibration certificate.