Learn More
Medchemexpress LLC HY-130233 5mg Medchemexpress, S516 CAS:1016543-77-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1302335MG
Encompass_Preferred
Medchemexpress, HY-130233 5mg S516 CAS:1016543-77-3 S516 (Compound 22) is an active metabolite of CKD-516 and a potent tubulin polymerization inhibitor with an IC50 of 4.29 μM. S516 has marked antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9823