Learn More
Medchemexpress LLC HY-117288 10mg Medchemexpress, S55746 CAS:1448584-12-0 Purity:>98%
SDP
Catalog No. 501969465
Encompass_Preferred
Medchemexpress, HY-117288 10mg S55746 CAS:1448584-12-0 S55746 (BLC201) is a potent, orally active and selective BCL-2 inhibitor, with a Ki of 1.3 nM and a Kd of 3.9 nM. S55746 (BLC201) has antitumor activity with low toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9465 Supplier Medchemexpress LLC Supplier No. HY11728810MG
Medchemexpress, HY-117288 10mg S55746 CAS:1448584-12-0 S55746 (BLC201) is a potent, orally active and selective BCL-2 inhibitor, with a Ki of 1.3 nM and a Kd of 3.9 nM. S55746 (BLC201) has antitumor activity with low toxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.