Learn More
Medchemexpress LLC S55746 hydrochloride 10mM 1mL | 1448525-91-4 | 1 ML
SDP
Catalog No. 5000227586
Encompass_Preferred
S55746 hydrochloride is a selective, orally active BCL-2 inhibitor supplied as a ready-to-use solution for in vitro research. The compound is provided at 10 mM in DMSO, with reported high purity and a molecular weight of approximately 747.3 g·mol⁻¹ (CAS 1448525-91-4).
  • Ready-to-use 10 mM solution in DMSO for convenience in assays.
  • High reported purity suitable for research applications.
  • Selective BCL-2 inhibition, useful in apoptosis and oncology studies.
  • Small-volume format minimizes waste for scarce compounds.
  • Supplied for research use only; not for human or clinical use.
Catalog No. 50-002-27586 Supplier Medchemexpress LLC Supplier No. HY117288A10MM 1ML
S55746 hydrochloride is a selective, orally active BCL-2 inhibitor supplied as a ready-to-use solution for in vitro research. The compound is provided at 10 mM in DMSO, with reported high purity and a molecular weight of approximately 747.3 g·mol⁻¹ (CAS 1448525-91-4).
  • Ready-to-use 10 mM solution in DMSO for convenience in assays.
  • High reported purity suitable for research applications.
  • Selective BCL-2 inhibition, useful in apoptosis and oncology studies.
  • Small-volume format minimizes waste for scarce compounds.
  • Supplied for research use only; not for human or clinical use.